Skip to Content
MilliporeSigma

79157

Fipronil-(pyrazole-13C3, cyano-13C)

PESTANAL®, analytical standard

Synonym(s):

5-Amino-3-cyano-13C-1-[2,6-dichloro-4-(trifluoromethyl)phenyl]- 4-[(trifluoromethyl)sulfinyl]pyrazole-13C3, Fipronil-13C4

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
13C4C8H4Cl2F6N4OS
Molecular Weight:
441.12
UNSPSC Code:
41116107
NACRES:
NA.24
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

product line

PESTANAL®

assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture

format

neat

storage temp.

2-8°C

SMILES string

N[13C]1=[13C](S(C(F)(F)F)=O)[13C]([13C]#N)=NN1C2=C(Cl)C=C(C(F)(F)F)C=C2Cl

InChI

1S/C12H4Cl2F6N4OS/c13-5-1-4(11(15,16)17)2-6(14)8(5)24-10(22)9(7(3-21)23-24)26(25)12(18,19)20/h1-2H,22H2/i3+1,7+1,9+1,10+1

InChI key

ZOCSXAVNDGMNBV-JOYIMARTSA-N

General description

Find more information here fipronil

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library