Skip to Content
MilliporeSigma

78447

Piperonylbutoxide

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

2-(2-Butoxyethoxy)ethyl (6-propylpiperonyl) ether, 4,5-Methylenedioxy-2-propylbenzyldiethyleneglycol butyl ether

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C19H30O5
CAS Number:
Molecular Weight:
338.44
EC Number:
200-076-7
UNSPSC Code:
41116107
NACRES:
NA.24
Beilstein/REAXYS Number:
288063
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

TraceCERT®

grade

certified reference material, TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

gas chromatography (GC): suitable

refractive index

n20/D 1.498 (lit.)

bp

155 °C/0.3 mmHg (lit.)

density

1.059 g/mL at 25 °C (lit.)

format

neat

storage temp.

2-8°C

SMILES string

CCCCOCCOCCOCc1cc2OCOc2cc1CCC

InChI

1S/C19H30O5/c1-3-5-7-20-8-9-21-10-11-22-14-17-13-19-18(23-15-24-19)12-16(17)6-4-2/h12-13H,3-11,14-15H2,1-2H3

InChI key

FIPWRIJSWJWJAI-UHFFFAOYSA-N

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Piperonylbutoxide


pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - STOT SE 3

target_organs

Respiratory system

supp_hazards

Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

339.8 °F - closed cup

flash_point_c

171 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library