Skip to Content
MilliporeSigma

77938

Methyl 3-hydroxyhexanoate

analytical standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CH3CH2CH2CH(OH)CH2CO2CH3
CAS Number:
Molecular Weight:
146.18
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
85151701
EC Number:
244-261-0
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

refractive index

n20/D 1.43 (lit.), n20/D 1.430

density

1 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCCC(O)CC(=O)OC

InChI

1S/C7H14O3/c1-3-4-6(8)5-7(9)10-2/h6,8H,3-5H2,1-2H3

InChI key

ACCRBMDJCPPJDX-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.


Still not finding the right product?

Explore all of our products under Methyl 3-hydroxyhexanoate


pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1

Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

185.0 °F - closed cup

flash_point_c

85 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library