Skip to Content
MilliporeSigma
All Photos(1)

Documents

77200

Sigma-Aldrich

tert-Butyl peroxybenzoate

technical, ≥95.0% (RT)

Synonym(s):

tert-Butyl perbenzoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COOOC(CH3)3
CAS Number:
Molecular Weight:
194.23
Beilstein:
1342734
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

6.7 (vs air)

Quality Level

vapor pressure

3.36 mmHg ( 50 °C)

grade

technical

Assay

≥95.0% (RT)

form

liquid

impurities

~2% tert-butyl benzoate

refractive index

n20/D 1.499 (lit.)
n20/D 1.50

bp

75-76 °C/0.2 mmHg (lit.)

density

1.021 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(C)(C)OOC(=O)c1ccccc1

InChI

1S/C11H14O3/c1-11(2,3)14-13-10(12)9-7-5-4-6-8-9/h4-8H,1-3H3

InChI key

GJBRNHKUVLOCEB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Sales restrictions may apply

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 3 - Org. Perox. C - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 2

Flash Point(F)

200.1 °F - closed cup - Decomposition

Flash Point(C)

93.4 °C - closed cup - Decomposition

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemical and Engineering News, 59, 19-19 (1981)
M Athar et al.
Carcinogenesis, 10(8), 1499-1503 (1989-08-01)
Humans are exposed to various peroxy and hydroperoxy compounds which are in use in the cosmetic, pharmaceutical and polymer industries and which are also generated as a result of the peroxidative metabolic conversion of certain lipids. This study was designed
Margaret Hanausek et al.
Carcinogenesis, 25(3), 431-437 (2003-11-25)
Screening of newly synthesized organic peroxides for tumor initiating/promoting activity would be greatly facilitated if predictive methodologies could be developed using topical exposures shorter than those required for definitive tumor assessment in mouse skin models. Nine organic peroxides [benzoyl peroxide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service