Skip to Content
MilliporeSigma

76847

Phosphate buffer solution

BioUltra, >0.044 M Na2HPO4, >0.022 M NaH2PO4

Synonym(s):

Phosphoric acid buffer solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
ND.02
UNSPSC Code:
12352200
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

BioUltra

Quality Segment

form

liquid

concentration

>0.022 M NaH2PO4, >0.044 M Na2HPO4

impurities

Insoluble matter, passes filter test, metal traces

pH

7.2±0.2

density

1.0 g/mL at 20 °C

absorption

0.02 at 260 nm, 0.02 at 280 nm

Application


  • Interfacial Engineering of MoS(2)@CoS(2) Heterostructure Electrocatalysts for Effective pH-Universal Hydrogen Evolution Reaction: Describes the application of phosphate buffer in creating stable electrochemical environments for the development of MoS(2)@CoS(2) heterostructure electrocatalysts, contributing to advancements in sustainable energy technologies (Gu YH et al., 2024).

  • Cryo-storage of porcine hides at the industrial scale for tissue engineering and regenerative medicine application: Investigates the use of phosphate buffer solutions to maintain pH stability during the cryopreservation of biological tissues, which is crucial for their viability in regenerative medicine and tissue engineering (Wang H et al., 2024).

  • Laser-printed paper ELISA and hydroxyapatite immobilization for colorimetric congenital anomalies screening in saliva: Highlights the application of phosphate buffer solutions in developing novel diagnostic platforms using laser-printed paper ELISA techniques, which could revolutionize prenatal screening practices by improving the accessibility and accuracy of testing for congenital anomalies (Moulahoum H et al., 2024).




Still not finding the right product?

Explore all of our products under Phosphate buffer solution


Storage Class

12 - Non Combustible Liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library