Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

75024

Supelco

3-Methoxytyramine hydrochloride

analytical standard

Synonym(s):

3-MT, 3-Methoxy-4-hydroxyphenethylamine hydrochloride, 4-(2-Aminoethyl)-2-methoxyphenol hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H3-4-(OH)CH2CH2NH2·HCl
CAS Number:
Molecular Weight:
203.67
Beilstein:
3631283
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥96.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤1.0% water

mp

213-215 °C (lit.)

application(s)

pharmaceutical

format

neat

storage temp.

−20°C

SMILES string

Cl[H].COc1cc(CCN)ccc1O

InChI

1S/C9H13NO2.ClH/c1-12-9-6-7(4-5-10)2-3-8(9)11;/h2-3,6,11H,4-5,10H2,1H3;1H

InChI key

AWRIOTVUTPLWLF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Methoxytyramine hydrochloride is a minor dopamine metabolite, found naturally in the cactus, Trichocereus pachanoi.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A new and highly sensitive method for measuring 3-methoxytyramine using HPLC with electrochemical detection. Studies with drugs which alter dopamine metabolism in the brain.
HealJ D, et al.
Neuropharmacology, 29(12), 1141-1150 (1990)
The Alkaloids (2007)
Edyta Możdżeń et al.
Neurotoxicity research, 27(3), 300-313 (2014-12-30)
Salsolinol (1-methyl-6,7-dihydroxy-1,2,3,4-tetrahydroisoquinoline), an endogenous compound present in the brain, was suspected of participation in the etiopathogenesis of Parkinson's disease, the most common serious movement disorder worldwide. In this study, we evaluated the effect of different (50, 100, and 500 µM)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service