Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

74567

Supelco

Permethrin-(phenoxy-d5)

mixture of cis and trans isomers, PESTANAL®, analytical standard

Synonym(s):

Permethrin-d5, [3-(Phenoxy-d5)phenyl]methyl 2,2-dimethyl-3-(2,2-dichlorovinyl)cyclopropanecarboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21D5H15Cl2O3
CAS Number:
Molecular Weight:
396.32
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

Assay

≥95.0% (sum of isomers, HPLC)

shelf life

limited shelf life, expiry date on the label

format

neat

SMILES string

ClC(=CC1C(C1C(=O)OCc2cc(ccc2)Oc3c(c(c(c(c3[2H])[2H])[2H])[2H])[2H])(C)C)Cl

InChI

1S/C21H20Cl2O3/c1-21(2)17(12-18(22)23)19(21)20(24)25-13-14-7-6-10-16(11-14)26-15-8-4-3-5-9-15/h3-12,17,19H,13H2,1-2H3/i3D,4D,5D,8D,9D

InChI key

RLLPVAHGXHCWKJ-YQYLVRRTSA-N

General description

Permethrin-(phenoxy-d5) is an isotope labeled analog of the pyrethroid insecticide permethrin, wherein the five phenoxy protons are replaced by deuterium.

Application

Isotope-labeled compounds are increasingly used in isotope dilution mass spectrometry (IDMS) for the quantitative analysis of pesticides. The major advantage of using this technique is that the isotope-labeled compounds have nearly the same physical properties as their non-labeled counterpart analogs, and thus show identical behavior during the workup and sample preparation process. This helps in overcoming the problems of matrix effects generally encountered in the usual LC-MS/GC-MS analysis, potentially resulting in biased results. By spiking the sample before workup with its isotope labeled analog, the loss of analyte that leads to matrix effects can be determined and compensated.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Microbial transformation of the pyrethroid insecticides: permethrin, deltamethrin, fastac, fenvalerate, and fluvalinate.
Maloney SE, et al.
Applied and Environmental Microbiology, 54(11), 2874-2876 (1988)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service