Skip to Content
MilliporeSigma

74346

Phenylpyruvic acid

analytical standard

Synonym(s):

2-Oxo-3-phenylpropanoic acid, 2-Oxo-3-phenylpropionic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H5CH2COCOOH
CAS Number:
Molecular Weight:
164.16
EC Number:
205-847-1
UNSPSC Code:
12352106
NACRES:
NA.24
Beilstein/REAXYS Number:
2207312
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

assay

≥97.0% (HPLC), 97.0-103.0% (T)

shelf life

limited shelf life, expiry date on the label

mp

150-154 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

−20°C

SMILES string

OC(=O)C(=O)Cc1ccccc1

InChI

1S/C9H8O3/c10-8(9(11)12)6-7-4-2-1-3-5-7/h1-5H,6H2,(H,11,12)

InChI key

BTNMPGBKDVTSJY-UHFFFAOYSA-N



Still not finding the right product?

Explore all of our products under Phenylpyruvic acid


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library