Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

74033

Supelco

Sudan II

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

1-(2,4-Xylylazo)-2-naphthol, Solvent Orange 7

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H16N2O
CAS Number:
Molecular Weight:
276.33
EC Number:
MDL number:
UNSPSC Code:
12171500
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

gas chromatography (GC): suitable

mp

156-158 °C (lit.)

SMILES string

Cc1ccc(\N=N\c2c(O)ccc3ccccc23)c(C)c1

InChI

1S/C18H16N2O/c1-12-7-9-16(13(2)11-12)19-20-18-15-6-4-3-5-14(15)8-10-17(18)21/h3-11,21H,1-2H3/b20-19+

InChI key

JBTHDAVBDKKSRW-FMQUCBEESA-N

Looking for similar products? Visit Product Comparison Guide

General description

Certified content by quantitative NMR incl. uncertainty and expiry are given in the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

448.9 °F - closed cup

Flash Point(C)

231.6 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service