Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

68694

Supelco

Imidacloprid

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10ClN5O2
CAS Number:
Molecular Weight:
255.66
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

[O-][N+](=O)NC1=NCCN1Cc2ccc(Cl)nc2

InChI

1S/C9H10ClN5O2/c10-8-2-1-7(5-12-8)6-14-4-3-11-9(14)13-15(16)17/h1-2,5H,3-4,6H2,(H,11,13)

InChI key

YWTYJOPNNQFBPC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Imidacloprid is a systemic chloronicotinyl insecticide, which can be effectively used in agriculture to control pests like aphids, scale insects, whiteflies, leafhoppers, some coleoptera and some lepidoptera species.

Find more information here: Neonicotinoids
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Application


  • Removal of neonicotinoids from wastewater: A study highlighted ferrate(VI)-based oxidation processes for removing neonicotinoids, including imidacloprid, from secondary effluents, suggesting a potential method for reducing environmental contamination from these chemicals (Real et al., 2024).

  • Resistance patterns in sheep blowflies: A study on in vitro insecticide resistance patterns in field strains of the sheep blowfly, Lucilia cuprina, included assessments of resistance to imidacloprid, providing insights into the development and spread of resistance in pest populations (Kotze et al., 2024).

  • Synergized insecticidal toxicity and risk reduction: The development of mesoporous silica nanospheres for the co-delivery of insecticides and antibiotics was studied to enhance the insecticidal efficacy of substances like imidacloprid while minimizing their environmental impact (Lv et al., 2024).

  • Alternative bio-insecticides against greenhouse pests: The efficacy of tangerine peel extracts as a bio-insecticide against Trialeurodes vaporariorum was evaluated, offering a potential alternative to chemical insecticides such as imidacloprid in agricultural pest control (Flores et al., 2024).

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Learn more about Neonicotinoids - active substances used in plant protection products to control harmful insects.

Learn more about Neonicotinoids - active substances used in plant protection products to control harmful insects.

Learn more about Neonicotinoids - active substances used in plant protection products to control harmful insects.

Learn more about Neonicotinoids - active substances used in plant protection products to control harmful insects.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service