Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

68450

Supelco

2-Methyl-1-pentene

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2C(CH3)=CH2
CAS Number:
Molecular Weight:
84.16
Beilstein:
1340499
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor pressure

325 mmHg ( 37.7 °C)

Assay

≥99.0% (GC)

autoignition temp.

572 °F

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.392 (lit.)
n20/D 1.392

bp

62 °C (lit.)

mp

−136 °C (lit.)

density

0.682 g/mL at 25 °C (lit.)

application(s)

environmental
petroleum

format

neat

SMILES string

CCCC(C)=C

InChI

1S/C6H12/c1-4-5-6(2)3/h2,4-5H2,1,3H3

InChI key

WWUVJRULCWHUSA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Methyl-1-pentene is a hydrocarbon belonging to the group of volatile organic compounds.

Application

2-Methyl-1-pentene may be used as an analytical reference standard for the quantification of the analyte in breath samples using gas chromatography technique.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-16.6 °F - closed cup

Flash Point(C)

-27 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The analysis of healthy volunteers' exhaled breath by the use of solid-phase microextraction and GC-MS
Ligor T, et al.
Journal of Breath Research, 2(4), 527-536 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service