Skip to Content
MilliporeSigma

67338

DL-2-Aminobutyric acid

analytical standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C2H5CH(NH2)CO2H
CAS Number:
Molecular Weight:
103.12
EC Number:
220-616-5
UNSPSC Code:
12352116
NACRES:
NA.24
Beilstein/REAXYS Number:
635889
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

assay

≥99% (TLC), 98.5-101.5% (NT)

form

solid

shelf life

limited shelf life, expiry date on the label

mp

291 °C (dec.) (lit.)

SMILES string

CCC(N)C(O)=O

InChI

1S/C4H9NO2/c1-2-3(5)4(6)7/h3H,2,5H2,1H3,(H,6,7)

InChI key

QWCKQJZIFLGMSD-UHFFFAOYSA-N



Still not finding the right product?

Explore all of our products under DL-2-Aminobutyric acid


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library