Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

66490

Supelco

Methylcyclopentane

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C5H9CH3
CAS Number:
Molecular Weight:
84.16
Beilstein:
1900214
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor pressure

232.8 mmHg ( 37.7 °C)

Assay

≥99.5% (GC)

autoignition temp.

624 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

8.35 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.409 (lit.)
n20/D 1.410

bp

72 °C (lit.)

mp

−142 °C (lit.)

density

0.748 g/mL at 20 °C
0.749 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

SMILES string

CC1CCCC1

InChI

1S/C6H12/c1-6-4-2-3-5-6/h6H,2-5H2,1H3

InChI key

GDOPTJXRTPNYNR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methylcyclopentane, a naphthene hydrocarbon, has a tertiary carbon atom and also has greater reactivity towards chemical agents like sulphuric acid, fuming nitric acid, etc.

Application

Methylcyclopentane was used to determine qualitative and quantitative interpretation of solvophobic effects in methyl cellosolve and propylene glycol, reading values of Gibbs free energy of solution and solvation.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

1.4 °F

Flash Point(C)

-17 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thermodynamics of solvation in propylene glycol and methyl cellosolve.
Sedov, I. A., M. A. Stolov, and B. N. Solomonov.
The Journal of Chemical Thermodynamics, 78, 32-36 (2014)
United states government printing office
Bureau of Standards Journal of Research, 7, 800-800 (1932)
Brian D Fitz et al.
Journal of chromatography. A, 1392, 82-90 (2015-03-31)
Low thermal mass gas chromatography (LTM-GC) was evaluated for rapid, high peak capacity separations with three injection methods: liquid, headspace solid phase micro-extraction (HS-SPME), and direct vapor. An Agilent LTM equipped with a short microbore capillary column was operated at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service