Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

63660

Supelco

(−)-Menthol

analytical standard

Synonym(s):

(1R,2S,5R)-2-Isopropyl-5-methylcyclohexanol, 5-Methyl-2-(1-methylethyl)cyclohexanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20O
CAS Number:
Molecular Weight:
156.27
Beilstein:
1902293
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.0% (sum of enantiomers, GC)

optical activity

[α]20/D −50±1°, c = 10% in ethanol

optical purity

enantiomeric ratio: ≥99:1 (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

~43 °C

application(s)

food and beverages

format

neat

SMILES string

CC(C)[C@@H]1CC[C@@H](C)C[C@H]1O

InChI

1S/C10H20O/c1-7(2)9-5-4-8(3)6-10(9)11/h7-11H,4-6H2,1-3H3/t8-,9+,10-/m1/s1

InChI key

NOOLISFMXDJSKH-KXUCPTDWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(-)-Menthol is a cyclic terpene alcohol that occurs in mentha species.It is responsible for their distinctive odor and flavor.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This compound is commonly found in plants of the genus: mentha
Important resolving agent for acids, recent applications include; Crystallization-induced asymmetric transformation of malonates ; Chiral auxiliary

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

201.2 °F - closed cup

Flash Point(C)

94 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Menthol: a refreshing look at this ancient compound
Patel T, et al.
Journal of the American Academy of Dermatology, 57(5), 873-878 (2007)
G. Solladie et al.
Synthesis, 173-173 (1987)
M. Ihara et al.
Journal of the Chemical Society. Chemical Communications, 9-9 (1988)
J.-M. Brunel et al.
The Journal of Organic Chemistry, 58, 7313-7313 (1993)
Ariane Leite Rozza et al.
PloS one, 9(1), e86686-e86686 (2014-01-28)
The aim of this research was to investigate the anti-apoptotic, antioxidant and anti-inflammatory properties of menthol against ethanol-induced gastric ulcers in rats. Wistar rats were orally treated with vehicle, carbenoxolone (100 mg/kg) or menthol (50 mg/kg) and then treated with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service