Skip to Content
MilliporeSigma
All Photos(1)

Documents

63011BC

Millipore

Supplement A, SimPlate®

BioControl, For use with SimPlate Total Plate Count and Yeast & Mold

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NA.85

material

plastic vial (containing dehydrated culture media)

form

liquid

shelf life

6‑36 mo.

manufacturer/tradename

BioControl Systems 63011-200

application(s)

food and beverages

compatibility

for use with SimPlate (66002BC)
for use with SimPlate (66003BC)
for use with SimPlate (66004BC)
for use with SimPlate (66007BC)
for use with SimPlate (66009BC)

storage temp.

2-8°C

suitability

molds
yeasts

General description

Supplement A is an additional reagent for use in the SimPlate® total plate count and yeast & mold. SimPlate® Binary Detection Technology® combines proprietary media and a patented plating device to provide fast and accurate quantitative results for key groups of indicator organisms. Defined substrates incorporated into the media are utilized to detect multiple enzymes and redox reactions associated with the target organisms resulting in chromogenic or fluorescent changes to the media, providing easy to read results. Supplement A is added to SimPlate® trays to alleviate the air bubbles in the media.

Application

Supplement A is added to SimPlate® trays to alleviate the air bubbles in media for the detection of pathogenic organisms in foods, ingredients, and environmental samples without the need for optimization of reaction and cycle conditions.

Components

1 bottle, approx. 22 mL (adequate for 200 tests)

Other Notes

For use with SimPlate (66002BC, 66003BC, 66004BC, 66007BC)

Legal Information

Binary Detection Technology is a registered trademark of Merck KGaA, Darmstadt, Germany
SimPlate is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service