Skip to Content
MilliporeSigma
All Photos(1)

Documents

61927

Supelco

Mesitylene

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

1,3,5-Trimethylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H3(CH3)3
CAS Number:
Molecular Weight:
120.19
Beilstein:
906806
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

vapor density

4.1 (vs air)

vapor pressure

14 mmHg ( 55 °C)
2.49 mmHg ( 25 °C)

product line

TraceCERT®

autoignition temp.

1022 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

0.88-6.1 %, 100 °F

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

storage condition

under inert gas (Argon)

technique(s)

gas chromatography (GC): suitable

refractive index

n20/D 1.499 (lit.)

bp

163-166 °C (lit.)

mp

−45 °C (lit.)

density

0.864 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

storage temp.

2-8°C

SMILES string

Cc1cc(C)cc(C)c1

InChI

1S/C9H12/c1-7-4-8(2)6-9(3)5-7/h4-6H,1-3H3

InChI key

AUHZEENZYGFFBQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

122.0 °F

Flash Point(C)

50 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service