Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

61827

Supelco

2,6-Bis{[bis(2-pyridylmethyl)amino]methyl}-4-methylphenol

for spectrophotometric determination of phosphate, ≥97.0% (HPLC)

Synonym(s):

H-bpmp

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H34N6O
CAS Number:
Molecular Weight:
530.66
Beilstein:
5321807
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.21

grade

for spectrophotometric determination of phosphate

Quality Level

Assay

≥97.0% (HPLC)

form

solid

technique(s)

UV/Vis spectroscopy: suitable

SMILES string

Cc1cc(CN(Cc2ccccn2)Cc3ccccn3)c(O)c(CN(Cc4ccccn4)Cc5ccccn5)c1

InChI

1S/C33H34N6O/c1-26-18-27(20-38(22-29-10-2-6-14-34-29)23-30-11-3-7-15-35-30)33(40)28(19-26)21-39(24-31-12-4-8-16-36-31)25-32-13-5-9-17-37-32/h2-19,40H,20-25H2,1H3

InChI key

SAJIFZPINIDYBV-UHFFFAOYSA-N

Application

Higly selective colorimetric dye for quantitve phosphate measurements in all kind of aqueous samples at pH 7.0. The indicator system is prepared by simply mixing H-bpmp, zinc perchlorate, and pyrocatechol violet in water in a 1:2:1 molar ratio.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Naked-eye detection of phosphate ions in water at physiological pH: a remarkably selective and easy-to-assemble colorimetric phosphate-sensing probe.
Min Su Han et al.
Angewandte Chemie (International ed. in English), 41(20), 3809-3811 (2002-10-19)
Synthesis and Properties of binuclear Cobalt (II) Oxygen Adduct with 2, 6-bis [bis (2-pyridylmethyl) aminomethyl]-4-methylphenol.
Suzuki, Masatatsu, Hajime Kanatomi, and Ichiro Murase.
Chemistry Letters (Jpn), 10 (12), 1745-1748 (1981)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service