Skip to Content
MilliporeSigma

60752

Silica gel Inorganic Sorbent

high-purity grade (w/ Ca, ~0.1%), 230-400 mesh

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
23201100
PubChem Substance ID:
NACRES:
SB.52
EC Number:
231-545-4
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Silica gel, high-purity grade (w/ Ca, ~0.1%), pore size 60 Å, 230-400 mesh particle size

grade

high-purity grade (w/ Ca, ~0.1%)

Quality Segment

description

activity acc. to Brockmann and Schodder 2 - 3, for flash chromatography, for preparative liquid chromatography

form

powder

manufacturer/tradename

Sigma-Aldrich

technique(s)

LPLC: suitable

surface area

500 m2/g

matrix

Silica

matrix active group

silica

particle size

230-400 mesh, 40-63 μm

pore size

0.8 cm3/g pore volume, 60 Å

bp

2230 °C

mp

>1600 °C

separation technique

hydrophilic interaction (HILIC)

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

General description

Silica gel is a rigid 3D network of colloidal silica. It is exclusively used as a support material for the active titanium (III) centers in Ziegler-Natta catalysts. Silica gel allows a controlled fragmentation leading to form uniform polymer particles with narrow particle size distribution and high bulk density. Silica gel is classified as below:
a. Aquagel - pores are filled with water
b. Xerogel - by the process of evaporation, aqueous phase in the pores are removed
c. Aerogel - solvent removed by supercritical extraction

Application

Silica gel may be used in separation of single-walled carbon nanotube (SWNT) by simple filtration.


Still not finding the right product?

Explore all of our products under Silica gel


Storage Class

13 - Non Combustible Solids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library