Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

59872

Supelco

2-[[2-[(2-Carboxybenzoyl)amino]ethoxy]methyl]-4-(2-chlorophenyl)-1,4-dihydro-6-methyl-3,5-pyridinedicarboxylic acid 3-ethyl 5-methyl ester

reference material

Synonym(s):

Amlodipine besilate impurity H (PhEur)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H29ClN2O8
CAS Number:
Molecular Weight:
556.99
MDL number:
UNSPSC Code:
41116107

grade

reference material

API family

amlodipine

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

format

neat

storage temp.

2-8°C

InChI

1S/C28H29ClN2O8/c1-4-39-28(36)24-21(15-38-14-13-30-25(32)17-9-5-6-10-18(17)26(33)34)31-16(2)22(27(35)37-3)23(24)19-11-7-8-12-20(19)29/h5-12,23,31H,4,13-15H2,1-3H3,(H,30,32)(H,33,34)

InChI key

IAYRABGXHCLELQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This reference material (RM) is produced and certified in accordance with ISO/IEC 17025. This RM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

related product

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1B - STOT RE 2 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service