Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

56919

Supelco

Perfluoroperhydrophenanthrene

Selectophore

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14F24
CAS Number:
Molecular Weight:
624.11
Beilstein:
2514226
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61
Pricing and availability is not currently available.

grade

for ion-selective electrodes

Quality Level

description

solvent for preparation of membranes

product line

Selectophore

bp

212-218 °C

density

2.03 g/mL at 20 °C

SMILES string

FC1(F)C(F)(F)C(F)(F)C2(F)C(F)(C1(F)F)C(F)(F)C(F)(F)C3(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C23F

InChI

1S/C14F24/c15-1-2(16)4(18,10(29,30)14(37,38)12(33,34)6(2,21)22)8(25,26)7(23,24)3(1,17)9(27,28)13(35,36)11(31,32)5(1,19)20

InChI key

QKENRHXGDUPTEM-UHFFFAOYSA-N

General description

Visit our Sensor Applications portal to learn more.

Application

Perfluoroperhydrophenanthrene finds application as an intraoperative and postoperative surgical adjunct tool, during retinal operations and study.[1][2]
Solvent for fluorous liquid phase in ion-selective electrodes

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

C Batman et al.
Ophthalmic surgery and lasers, 29(2), 144-146 (1998-03-21)
Perfluorocarbon liquids have gained wide acceptance as intraoperative tools that can simplify vitreoretinal surgical maneuvers. These low-viscosity liquids facilitate injection into the eye and removal from the eye during surgery. Tolerance to perfluoroperhydrophenanthrene and the development of proliferative vitreoretinopathy with
U Stolba et al.
Retina (Philadelphia, Pa.), 17(2), 146-153 (1997-01-01)
Anterior and posterior segment changes of experimental vitreous and aqueous substitution with Perfluorophenanthrene were evaluated. In 28 rabbit eyes that underwent vitrectomy, tamponades of 1.2 cc Perfluorophenanthrene remained as long as 8 weeks under clinical and electrophysiologic control. Histologic examinations
Paul G Boswell et al.
Journal of the American Chemical Society, 127(25), 8958-8959 (2005-06-23)
Potentiometric sensors based on fluorous membranes doped with a fluorophilic tetraphenylborate derivative are shown to have a remarkably wide range of selectivities that exceeds the selectivity range of conventional polymeric membranes by 8 orders of magnitude. The fluorous character of
Paul G Boswell et al.
Journal of the American Chemical Society, 127(48), 16976-16984 (2005-12-01)
Despite the widespread use of perfluorinated solvents with amino and ether groups in a variety of application fields, the coordinative properties of these compounds are poorly known. It is generally assumed that the electron withdrawing perfluorinated moieties render these functional
Chun-Ze Lai et al.
Analytical chemistry, 82(18), 7634-7640 (2010-08-31)
Ionophore-doped sensing membranes exhibit greater selectivities and wider measuring ranges if their membrane matrixes are noncoordinating and solvate interfering ions poorly. This is particularly true for fluorous phases, which are the least polar and polarizable condensed phases known. In this

Questions

  1. What is the percentage purity of this product?

    1 answer
    1. Unfortunately, this product is not tested for purity.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service