Skip to Content
MilliporeSigma

56480

N-Hydroxysuccinimide

≥97.0% (T), for peptide synthesis

Synonym(s):

1-Hydroxy-2,5-pyrrolidinedione, HOSu, NHS

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C4H5NO3
CAS Number:
Molecular Weight:
115.09
NACRES:
NA.22
PubChem Substance ID:
UNSPSC Code:
12352005
EC Number:
228-001-3
MDL number:
Beilstein/REAXYS Number:
113913
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

N-Hydroxysuccinimide, purum, ≥97.0% (T)

grade

purum

Quality Segment

assay

≥97.0% (T)

form

solid

reaction suitability

reaction type: Addition Reactions

mp

95-98 °C

application(s)

peptide synthesis

functional group

imide

SMILES string

ON1C(=O)CCC1=O

InChI

1S/C4H5NO3/c6-3-1-2-4(7)5(3)8/h8H,1-2H2

InChI key

NQTADLQHYWFPDB-UHFFFAOYSA-N

Application

N-Hydroxysuccinimide (NHS) can be used in the following applications:
  • To synthesize N-succinimidyl 3-(2-pyridyldithio)-propionate, a heterobifunctional reagent useful for protein-protein conjugation and also to incorporate aliphatic thiols into proteins.
  • To synthesize NHS esters of long-chain fatty acids.
  • NHS can activate the phosphonic acid monolayers immobilized on titanium surface for binding with proteins.

Other Notes

may contain 1-3% succinic acid and/or succinic anhydride


Still not finding the right product?

Explore all of our products under N-Hydroxysuccinimide


pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library