Skip to Content

52017

3-Hydroxybutyric acid

analytical standard

Synonym(s):

(±)-3-Hydroxybutanoic acid, DL-β-Hydroxybutyric acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CH3CH(OH)CH2COOH
CAS Number:
Molecular Weight:
104.10
EC Number:
206-099-9
UNSPSC Code:
41116107
NACRES:
NA.24
Beilstein/REAXYS Number:
773861
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.4390-1.4450, n20/D 1.443 (lit.)

bp

118-120 °C/2 mmHg (lit.)

density

1.126 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(O)CC(O)=O

InChI

1S/C4H8O3/c1-3(5)2-4(6)7/h3,5H,2H2,1H3,(H,6,7)

InChI key

WHBMMWSBFZVSSR-UHFFFAOYSA-N



Still not finding the right product?

Explore all of our products under 3-Hydroxybutyric acid


Pictograms

Corrosion

Signal Word

Danger

Hazard Codes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

Flash Point (°F)

233.6 °F - closed cup

Flash Point (°C)

112 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library