Skip to Content
MilliporeSigma

51971

Zirconium Standard for ICP

TraceCERT®, 10 g/L Zr in nitric acid and hydrofluoric acid (nominal concentration)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

TraceCERT®

grade

certified reference material, TraceCERT®

concentration

10 g/L Zr in nitric acid and hydrofluoric acid (nominal concentration)

technique(s)

ICP: suitable

application(s)

industrial qc
pharmaceutical

format

single component solution

General description

This certified reference material (CRM) is produced in accordance with ISO 17034 and characterized in accordance with ISO/IEC 17025. This CRM is traceable to SI unit kg and measured against primary material from a National Metrology Institute (NMI), e.g. NIST.

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.

Search & download your certificate here.

Application

Zirconium standard can be used as a calibration standard in the determination ofzirconium in surface water samples by single particle technique with argonplasma-coupled mass spectrometry (SP-ICP-MS)

Analysis Note

10 g/L Zr in 5% nitric acid and <0.5% hydrofluoric acid, prepared with high purity Zr metal, HNO3, HF and water

Other Notes

For a complete product listing of our ICP and AAS range, including technical information and example certificates, please visit ICP & AAS standards website.

Detailed information on our complete workflow solutions for ICP-AAS can be found in our Brochure: High-Purity Chemicals for Inorganic Trace Analysis.

In our effort to be more sustainable, we′re going digital with COAs-download them directly from our website.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Zirconium Standard for ICP


pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

supp_hazards

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library