Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

51602

Supelco

Sudan Red B

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H20N4O
CAS Number:
Molecular Weight:
380.44
Colour Index Number:
26110
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

173-175 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

Cc1cccc(c1)\N=N\c2ccc(cc2C)\N=N\c3c(O)ccc4ccccc34

InChI

1S/C24H20N4O/c1-16-6-5-8-19(14-16)25-27-22-12-11-20(15-17(22)2)26-28-24-21-9-4-3-7-18(21)10-13-23(24)29/h3-15,29H,1-2H3/b27-25+,28-26+

InChI key

NHXXLZBKTKNTEF-NBHCHVEOSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service