Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

49894

Supelco

Sodium propionate

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

Propionic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2COONa
CAS Number:
Molecular Weight:
96.06
Beilstein:
3566934
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
E Number:
E281
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

285-286 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

[Na+].CCC([O-])=O

InChI

1S/C3H6O2.Na/c1-2-3(4)5;/h2H2,1H3,(H,4,5);/q;+1/p-1

InChI key

JXKPEJDQGNYQSM-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Bistra B Nankova et al.
PloS one, 9(8), e103740-e103740 (2014-08-30)
Alterations in gut microbiome composition have an emerging role in health and disease including brain function and behavior. Short chain fatty acids (SCFA) like propionic (PPA), and butyric acid (BA), which are present in diet and are fermentation products of
Kelly A Foley et al.
Behavioural brain research, 278, 244-256 (2014-10-11)
Potential environmental risk factors for autism spectrum disorders (ASD) include viral/bacterial infection and an altered microbiome composition. The present study investigated whether administration of immune and gastrointestinal factors during gestation and early life altered startle response and prepulse inhibition in
Guojun Shi et al.
Journal of molecular endocrinology, 53(3), 367-380 (2014-10-10)
Recent reports have highlighted the roles of free fatty acid receptor 2 (FFAR2) in the regulation of metabolic and inflammatory processes. However, the potential function of FFAR2 in type 1 diabetes (T1D) remains unexplored. Our results indicated that the mRNA
Genlai Li et al.
The Journal of nutrition, 144(12), 1887-1895 (2014-10-17)
Short-chain fatty acids (SCFAs), including acetate, propionate, and butyrate, are the main products of microbial fermentation in the gut and might mediate some of the effects of gut microbiota and nutrition on development, metabolism, and pathogenesis of obesity and other
Rémi Legay et al.
Journal of pharmaceutical and biomedical analysis, 98, 446-462 (2014-07-20)
The purpose of the study was to investigate the degradation pathway of 5-fluorouracil (FU) in the situation of commercial formulations for clinical use, namely FU dissolved in sodium hydroxide (NaOH) solutions or Tris buffer at pH 8.5-9. Combination of data

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service