Skip to Content
MilliporeSigma
All Photos(1)

Documents

49722

Supelco

4-Methyl-3-penten-2-one

analytical standard

Synonym(s):

Mesityl oxide, 4-Methyl-3-penten-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHCOCH3
CAS Number:
Molecular Weight:
98.14
Beilstein:
1361550
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.5% water

refractive index

n20/D 1.440-1.446
n20/D 1.442 (lit.)

bp

130 °C (lit.)

mp

-41.5 °C (lit.)

density

0.858 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

C\C(C)=C\C(C)=O

InChI

1S/C6H10O/c1-5(2)4-6(3)7/h4H,1-3H3

InChI key

SHOJXDKTYKFBRD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Methyl-3-penten-2-one belongs to the volatile organic compounds (VOCs) class of chemicals.

Application

4-Methyl-3-penten-2-one may be used as a reference standard for the determination of 4-methyl-3-penten-2-one in Irish beef meats by gas chromatography-olfactometry (GC-O), in human urine by static headspace extraction followed by analysis using GC with mass spectrometry (MS), in caja-umbu (Spondias sp.) fruits by GC-MS and in wild gilthead sea bream (Sparus aurata) by simultaneous distillation-extraction (SDE) followed by analysis using GC with flame ionization detector (FID) as well as GC-MS.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

87.8 °F - closed cup

Flash Point(C)

31 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A comparative study of the analysis of human urine headspace using gas chromatography-mass spectrometry.
Smith S, et al.
Journal of Breath Research, 2(3), 037022-037022 (2008)
Gas chromatography-olfactometry analysis of the volatile compounds of two commercial Irish beef meats.
Machiels D, et al.
Talanta, 60(4), 755-764 (2003)
Analysis of volatile compounds of wild gilthead sea bream (Sparus aurata) by simultaneous distillation-extraction (SDE) and GC-MS.
Selli S and Cayhan GG
Microchemical Journal, Devoted to the Application of Microtechniques in All Branches of Science, 93(2), 232-235 (2009)
Volatile compounds captured through purge and trap technique in caja-umbu (Spondias sp.) fruits during maturation.
Narain N, et al.
Food Chemistry, 102(3), 726-731 (2007)
L J Rose et al.
Current microbiology, 41(3), 206-209 (2000-07-29)
Volatile organic compounds from Penicillium viridicatum and Methylobacterium mesophilicum growing on laboratory media and on component materials of automobile air conditioners were analyzed with gas chromatography and mass spectrometry. P. viridicatum produced compounds such as 4-methyl thiazole, terpenes and alcohols

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service