Skip to Content
MilliporeSigma

48121

ProClin 150

ProClin preservative for use in IVD manufacturing, CMIT/MIT 1.5%, Method Development Kit, pkg of 3 × 5 mL each (three different lots)

Synonym(s):

1.5% solution of 5-chloro-2-methyl-4-isothiazolin-3-one and 2-Methyl-4-isothiazolin-3-one

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352201
NACRES:
NA.28
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

description

biocide for IVD use

form

liquid

shelf life

2 yr (from Date of Manufacture)

contains

23 to 25% Mg salts

composition

CMIT/MIT, 1.5%

packaging

pkg of 3 × 5 mL each (three different lots)

parameter

operating temperature at <45 °C

matrix

Water

operating pH range

2.5-8.5

pH range

1.7-3.7

bp

100 °C/1013 hPa

mp

-21 °C

solubility

water: soluble

density

1.20 g/cm3

Application

ProClin preservatives are immediately microstatic upon contact with microbial organisms as a result of their ability to quickly penetrate cell membranes and inhibit specific enzymes crucial to cellular respiration. Multiple specific sites of ProClin toxicity prevent microorganisms from developing high levels of resistance, making ProClin highly efficient even over extended storage periods of your IVD components.

Legal Information

ProClin is a trademark of LANXESS Corp.

Disclaimer

The product is not intended for use as a biocide under global biocide regulations, including but not limited to US EPA′s Federal Insecticide Fungicide and Rodenticide Act, European Biocidal Products Regulation, Canada′s Pest Management Regulatory Agency, Turkey′s Biocidal Products Regulation, Korea′s Consumer Chemical Products and Biocide Safety Management Act (K-BPR) and others.


Still not finding the right product?

Explore all of our products under ProClin 150


signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library