Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

45631

Supelco

Prochloraz

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H16Cl3N3O2
CAS Number:
Molecular Weight:
376.67
Beilstein:
8155344
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCCN(CCOc1c(Cl)cc(Cl)cc1Cl)C(=O)n2ccnc2

InChI

1S/C15H16Cl3N3O2/c1-2-4-20(15(22)21-5-3-19-10-21)6-7-23-14-12(17)8-11(16)9-13(14)18/h3,5,8-10H,2,4,6-7H2,1H3

InChI key

TVLSRXXIMLFWEO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Prochloraz belongs to the imidazole class of fungicides, with protective and curative action, found to be effective against different fungi affecting cereal crops, turf, citrus and tropical fruit, field legumes and beet. Its mode of action involves the inhibition of ergosterol biosynthesis.

Application

Prochloraz may be used as an analytical reference standard for the quantification of the analyte in agricultural commodities using different chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simplified determination of combined residues of prochloraz and its metabolites in vegetable, fruit and wheat samples by gas chromatography
Paoli D.M, et al.
Journal of Chromatography A, 765(1), 127-131 (1997)
Nigel C Noriega et al.
Biology of reproduction, 72(6), 1324-1335 (2005-01-28)
Prochloraz (PZ) is an imidazole fungicide that displays multiple endocrine activities. It inhibits steroid synthesis via P450 modulation and acts as an androgen receptor (AR) antagonist, but its effects on male sexual differentiation have not been described. The purpose of
The Pesticide Encyclopedia (2014)
Determination of fungicide residues in fruits and vegetables by liquid chromatography-atmospheric pressure chemical ionization mass spectrometry
Blasco C, et al.
Journal of Chromatography A, 947(2), 227-235 (2002)
Karen L Thorpe et al.
Aquatic toxicology (Amsterdam, Netherlands), 105(3-4), 412-420 (2011-08-13)
Exogenous treatment of fish with natural sex hormones and their mimics has been shown to influence gonadal differentiation resulting in biased phenotypic sex-ratios. This has lead to the development of the Fish Sexual Development Test (FSDT) as a method for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service