Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

44770

Supelco

Mettler-Toledo Calibration substance ME 30034252, Phenyl salicylate

traceable to primary standards

Synonym(s):

Phenyl salicylate, Salol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(HO)C6H4CO2C6H5
CAS Number:
Molecular Weight:
214.22
Beilstein:
393969
EC Number:
MDL number:
UNSPSC Code:
20121904
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

bp

172-173 °C/12 mmHg (lit.)

mp

41-43 °C (lit.)

application(s)

food and beverages
pharmaceutical

format

neat

SMILES string

Oc1ccccc1C(=O)Oc2ccccc2

InChI

1S/C13H10O3/c14-12-9-5-4-8-11(12)13(15)16-10-6-2-1-3-7-10/h1-9,14H

InChI key

ZQBAKBUEJOMQEX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Phenyl salicylate is a carboxylic acid phenyl ester, known for its aroma and flavoring properties. It finds application in foods and consumer products, medicines and as a UV filter.

Mettler-Toledo calibration substance ME 30034252, phenyl salicylate is an analytical standard, intended for use in the regular calibration of Mettler-Toledo melting point instrument. Its melting point value is an average of 6 to 12 iterations with a Mettler-Toledo MP90 Excellence instrument that is calibrated against primary standards. The melting point is measured using Capillary method in conformance with European Pharmacopeia (2.2.14.)

Application

Vanillin Mettler-Toledo calibration substance ME 51143093 is used to calibrate Mettler-Toledo melting point instruments.

Features and Benefits

  • Available in a ready-to-use bottle as a finely ground powder
  • Melting point calibration standard traceable to a primary standard (LGC, London)
  • Melting point evaluation conducted in both thermodynamic and pharmacopeia modes for physically correct and heating rate dependent melting point determinations, respectively
  • Provided with certificates of analysis and safety sata sheet
  • A product of analytical standard grade to help meet the QC/QA requirements of melting point determination
  • Standard deviation up to ± 0.2 °C

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of the UV-filters benzyl salicylate, phenyl salicylate, octyl salicylate, homosalate, 3-(4-methylbenzylidene) camphor and 3-benzylidene camphor in human placental tissue by LC-MS/MS. Assessment of their in vitro endocrine activity
Jimenez-Diaz I, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 80-87 null
Thermodynamic studies on the interactions of phenyl salicylate in protic solvents at different temperatures
Iqbal MJ and Chaudhry MA
Journal of Molecular Liquids, 75-80 null
Hydrolytic metabolism of phenyl and benzyl salicylates, fragrances and flavoring agents in foods, by microsomes of rat and human tissues
Ozaki H, et al.
Food And Chemical Toxicology, 116-123 null
Cross-sensitivity between resorcinol, resorcinol monobenzoate and phenyl salicylate.
M Nakagawa et al.
Contact dermatitis, 27(3), 199-200 (1992-09-01)
C D Calnan et al.
Contact dermatitis, 7(4), 208-211 (1981-07-01)
Six cases of contact dermatitis from a lip salve are described. Five were allergic to phenyl salicylate and one to geraniol in the fragrance. The dermatitis spread in a ring around the mouth. Phenyl salicylate has been removed from the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service