Skip to Content
MilliporeSigma
All Photos(1)

Documents

4465962

Supelco

Amplifex Keto Reagent Kit

Synonym(s):

Ketone Body Quantification Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000
NACRES:
NA.21

technique(s)

LC/MS: suitable

shipped in

dry ice

storage temp.

−20°C

General description

Kit contains a protocol, 2 tubes of Amplifex Keto reagent and 2 tubes of diluent.  Each tube contains enough reagent to complete 25 reactions.

Application

It was used in targeted metabolomic analysis, in a study done to understand the mitochondrial thiol modification, by targeted electrophile, which inhibits metabolism in breast adenocarcinoma cells. It may also be used in the oxosteroid chemical derivatization kit, in a study to determine the neurosteroids in sea lamprey by UPLC-MS/MS.

Analysis Note

Additional information about the Amplifex Keto Reagent kit is at this page.

To view the Protocol for the Amplifex Keto Reagent Kit, please visit this Reagent protocol.

Legal Information

Amplifex is a trademark of AB Sciex Pte. Ltd.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ugo Bussy et al.
Talanta, 149, 326-334 (2016-01-01)
This article describes the development and validation of a sensitive and robust method for the determination of neurosteroids in sea lamprey, an ancestral animal in vertebrate evolution. Chemical derivatization was used to enhance the detection of neurosteroids containing ketone function
M Ryan Smith et al.
Redox biology, 8, 136-148 (2016-01-18)
Many cancer cells follow an aberrant metabolic program to maintain energy for rapid cell proliferation. Metabolic reprogramming often involves the upregulation of glutaminolysis to generate reducing equivalents for the electron transport chain and amino acids for protein synthesis. Critical enzymes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service