Skip to Content
MilliporeSigma
All Photos(1)

Documents

44208

Supelco

L-Lysine hydrochloride solution

100 mM amino acid in 0.1 M HCl, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14N2O2 · HCl
CAS Number:
Molecular Weight:
182.65
Beilstein:
5460909
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

form

liquid

analyte chemical class(es)

amino acids, peptides, proteins

concentration

100 mM amino acid in 0.1 M HCl

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

color

colorless

application(s)

food and beverages

format

single component solution

storage temp.

2-8°C

SMILES string

Cl.NCCCC[C@H](N)C(O)=O

InChI

1S/C6H14N2O2.ClH/c7-4-2-1-3-5(8)6(9)10;/h5H,1-4,7-8H2,(H,9,10);1H/t5-;/m0./s1

InChI key

BVHLGVCQOALMSV-JEDNCBNOSA-N

General description

L-Lysine hydrochloride is one of the essential amino acids which usually finds applications in the treatment of shingles and genital as well mouth lesions attributed to herpes zoster and herpes simplex virus, respectively.

Application

L-Lysine hydrochloride may be used as a reference standard for the determination of L-lysine hydrochloride in pharmaceutical dosage form by high resolution high performance liquid chromatography (HR-HPLC). It may be used as a precursor of lysine for its determination in breath condensate by isotope dilution hydrophilic-interaction liquid chromatography combined with tandem mass spectrometry (HILIC-MS/MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

nwg

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development and validation of new hplc method for simultaneous estimation of L-lysine hydrochloride and L-carnitine-L-tartrate in pharmaceutical dosage form.
Qadir MA, et al.
Indian Journal of Pharmaceutical Sciences, 77(4), 434-434 (2015)
Simultaneous determination of the advanced glycation end product Nε-carboxymethyllysine and its precursor, lysine, in exhaled breath condensate using isotope-dilution-hydrophilic-interaction liquid chromatography coupled to tandem mass spectrometry
Schettgen T, et al.
Analytical and Bioanalytical Chemistry, 387(8), 2783-2791 (2007)
Guntur Fibriansah et al.
Nature communications, 6, 6341-6341 (2015-02-24)
Dengue virus (DENV) infects ~400 million people annually. There is no licensed vaccine or therapeutic drug. Only a small fraction of the total DENV-specific antibodies in a naturally occurring dengue infection consists of highly neutralizing antibodies. Here we show that
Anne M Muskalla et al.
Clinical chemistry, 60(12), 1543-1548 (2014-10-11)
G-protein receptor kinase 4 polymorphism influences blood pressure regulation via modulation of dopamine receptor D1 in renal proximal tubular cells. We investigated the role of G-protein receptor kinase 4 polymorphism in the response to hypertensive therapy in patients with essential
Rick Kapur et al.
British journal of haematology, 166(6), 936-945 (2014-06-10)
Haemolytic disease of the fetus and newborn (HDFN) may occur when maternal IgG antibodies against red blood cells (RBCs), often anti-RhD (anti-D) antibodies, cross the placenta and mediate the destruction of RBCs via phagocytic IgG-Fc-receptors (FcγR). Clinical severity is not

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service