Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

44189

Supelco

Isoamyl 4-methoxycinnamate

analytical standard

Synonym(s):

Isoamyl p-methoxycinnamate, 4-Methoxycinnamic acid 3-methylbutyl ester, 4-Methoxycinnamic acid isoamyl ester, Amiloxate, Isopentyl 4-methoxycinnamate, NSC 408332

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H20O3
CAS Number:
Molecular Weight:
248.32
Beilstein:
3132627
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

O(C)c1ccc(cc1)\C=C\C(=O)OCCC(C)C

InChI

1S/C15H20O3/c1-12(2)10-11-18-15(16)9-6-13-4-7-14(17-3)8-5-13/h4-9,12H,10-11H2,1-3H3/b9-6+

InChI key

UBNYRXMKIIGMKK-RMKNXTFCSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Isoamyl 4-methoxycinnamate is a derivative of cinnamon acid ester and may be used as a sunscreen agent since it acts as a waterproof filter for ultraviolet B (UVB) rays.

Application

Isoamyl 4-methoxycinnamate may be used as a reference standard in the determination of isoamyl 4-methoxycinnamate in water samples using liquid chromatography coupled with ultraviolet spectrophotometry detector (LC-UV).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F W Jekat et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 30(9), 803-807 (1992-09-01)
3-(4-methoxyphenyl)-3-methylbutylester, propenoic acid (MPMBE; CAS no. 71617-10-2), a UVB-light filter used for sun protection of the skin, was administered once daily by intragastric gavage to pregnant Wistar rats on days 6-15 of gestation. Doses of 0.25 ml/kg/day (study group: D0.25)
Ionic liquid-based single-drop microextraction followed by liquid chromatography-ultraviolet spectrophotometry detection to determine typical UV filters in surface water samples.
Vidal L, et al.
Talanta, 81(1-2), 549-555 (2010)
Photo-allergic contact dermatitis caused by isoamyl p-methoxycinnamate in an 'organic' sunscreen.
Mohammad K Ghazavi et al.
Contact dermatitis, 64(2), 115-116 (2011-01-08)
Year Book of Dermatology and Dermatological Surgery 2012 - E-Book (2012)
Han-Shen Zhen et al.
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials, 31(5), 677-678 (2008-10-02)
To analyze the volatile oils chemical constituents of roots of Actinidia deliciosa. The volatile oils fraction of roots of Actinidia deliciosa. were extracted by water vapor distilling, and then the constituents were separated and identified, by GC-MS. 16 compounds were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service