Skip to Content
MilliporeSigma
All Photos(1)

Documents

42598

Supelco

Pullulan Standard

certified reference material, certified according to DIN, for GPC, 22,000

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41116107
E Number:
E1204
NACRES:
NA.24

grade

certified reference material
for GPC

Quality Level

shelf life

limited shelf life, expiry date on the label

mol wt

Mn ~21300
Mp ~22000
Mw ~22800
Mw/Mn ~1.07

analyte chemical class(es)

oligosaccharides

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Pullulan Standards are mostly used to obtain molecular mass distribution of cellulosic samples. It mainly consists of polymaltotriose units linked together by α-(1→6) linkages.

Application

It may be used in determination of molecular weight of hydrolysate using gel permeation chromatography, in a study performed to investigate enzymatic degradation of chitins and partially deacetlylated chitins.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enzymatic degradation of chitins and partially deacetylated chitins.
Shigemasa, Yoshihiro, et al.
International Journal of Biological Macromolecules, 16.1, 43-49 (1994)
Chi-San Wu
Handbook Of Size Exclusion Chromatography And Related Techniques: Revised And Expanded, 336-336 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service