Skip to Content

42446

Bis(8-methylnonyl) phthalate

analytical standard

Synonym(s):

1,2-Benzenedicarboxylic acid 1,2-bis(8-methylnonyl) ester, Diisodecyl phthalate, Phthalic acid bis(8-methylnonyl) ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C28H46O4
CAS Number:
Molecular Weight:
446.66
PubChem Substance ID:
UNSPSC Code:
85151701
NACRES:
NA.24
EC Number:
201-884-2
MDL number:
Beilstein/REAXYS Number:
2171889
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

refractive index

n20/D 1.477-1.481

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

O(CCCCCCCC(C)C)C(=O)c1c(cccc1)C(=O)OCCCCCCCC(C)C

InChI

1S/C28H46O4/c1-23(2)17-11-7-5-9-15-21-31-27(29)25-19-13-14-20-26(25)28(30)32-22-16-10-6-8-12-18-24(3)4/h13-14,19-20,23-24H,5-12,15-18,21-22H2,1-4H3

InChI key

ZVFDTKUVRCTHQE-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.


Storage Class

10 - Combustible liquids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library