Skip to Content
MilliporeSigma

Skip To

42272

Valeraldehyde

analytical standard

Synonym(s):

Pentanal

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
CH3(CH2)3CHO
CAS Number:
Molecular Weight:
86.13
UNSPSC Code:
85151701
NACRES:
NA.24
PubChem Substance ID:
EC Number:
203-784-4
Beilstein/REAXYS Number:
1616304
MDL number:
eCl@ss:
39021107
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

assay

≥97.5% (GC)

autoignition temp.

428 °F

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

impurities

≤0.5% water

refractive index

n20/D 1.394 (lit.)

bp

102-103 °C (lit.)

mp

−92 °C (lit.)

density

0.81 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

[H]C(=O)CCCC

InChI

1S/C5H10O/c1-2-3-4-5-6/h5H,2-4H2,1H3

InChI key

HGBOYTHUEUWSSQ-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
616964321774103
technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

format

neat

format

neat

format

neat

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

grade

analytical standard

grade

analytical standard

grade

analytical standard

grade

analytical standard

assay

≥97.5% (GC)

assay

≥97.0% (GC)

assay

≥97.0% (GC)

assay

≥99.0% (GC)


Still not finding the right product?

Explore all of our products under Valeraldehyde


pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

44.6 °F

flash_point_c

7 °C



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters