Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

41694

Supelco

5-Hydroxydiclofenac

analytical standard

Synonym(s):

5-OH-DCF, 2-[(2,6-Dichlorophenyl)amino]-5-hydroxybenzeneacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H11Cl2NO3
CAS Number:
Molecular Weight:
312.15
Beilstein:
4199419
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical

format

neat

storage temp.

−20°C

SMILES string

OC(CC1=CC(O)=CC=C1NC2=C(Cl)C=CC=C2Cl)=O

InChI

1S/C14H11Cl2NO3/c15-10-2-1-3-11(16)14(10)17-12-5-4-9(18)6-8(12)7-13(19)20/h1-6,17-18H,7H2,(H,19,20)

InChI key

VNQURRWYKFZKJZ-UHFFFAOYSA-N

General description

5-Hydroxydiclofenac, a monohydroxylated metabolite of diclofenac, belongs to the class of non-steroidal anti-inflammatory compounds.29}

Application

5-Hydroxydiclofenac may be used as an analytical standard for the determination of the analyte in biological samples and wastewater by liquid chromatography (LC) based techniques.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jing Shi et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 191, 1-7 (2017-10-02)
A novel flow injection chemiluminescence immunoassay for simple, sensitive and low-cost detection of diclofenac was established based on specific binding of antigen and antibody. Carboxylic resin beads used as solid phase carrier materials provided good biocompatibility and large surface-to-volume ratio
High-performance liquid chromatographic determination of diclofenac and its monohydroxylated metabolites in biological fluids
Godbillon J, et al.
Journal of Chromatography. B, Biomedical Applications, 338(1), 151-159 (1985)
High-performance liquid chromatographic method for the routine determination of diclofenac and its hydroxy and methoxy metabolites from in vitro systems
Schmitz G, et al.
Journal of Chromatography. B, Biomedical Applications, 620(1), 158-163 (1993)
Analysis of diclofenac and four of its metabolites in human urine by HPLC
Sawchuk RJ, et al.
Pharmaceutical Research, 12(5), 756-762 (1995)
Simultaneous determination of diclofenac, its human metabolites and microbial nitration/nitrosation transformation products in wastewaters by liquid chromatography/quadrupole-linear ion trap mass spectrometry
Osorio V, et al.
Journal of Chromatography A, 1347(1), 63-71 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service