Skip to Content
MilliporeSigma

41260

Bis(4-methylpentyl)phthalate

analytical standard

Synonym(s):

Phthalic acid bis(4-methylpentyl) ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H30O4
CAS Number:
Molecular Weight:
334.45
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
77101502
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

assay

≥95.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

refractive index

n20/D 1.484-1.488

application(s)

environmental

format

neat

storage temp.

2-8°C

SMILES string

O=C(OCCCC(C)C)C1=CC=CC=C1C(OCCCC(C)C)=O

InChI

1S/C20H30O4/c1-15(2)9-7-13-23-19(21)17-11-5-6-12-18(17)20(22)24-14-8-10-16(3)4/h5-6,11-12,15-16H,7-10,13-14H2,1-4H3

InChI key

ALEROMXYYSQFLX-UHFFFAOYSA-N



pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Repr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library