Skip to Content
MilliporeSigma

Skip To

39784

Isopentenyl pyrophosphate trilithium salt

analytical standard

Synonym(s):

3-Methyl-3-butenyl pyrophosphate trilithium salt, IPP

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C5H9Li3O7P2
CAS Number:
Molecular Weight:
263.89
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
85151701
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

grade

analytical standard

assay

≥95.0% (TLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

SMILES string

[Li+].[Li+].[Li+].CC(=C)CCOP([O-])(=O)OP([O-])([O-])=O

InChI

1S/C5H12O7P2.3Li/c1-5(2)3-4-11-14(9,10)12-13(6,7)8;;;/h1,3-4H2,2H3,(H,9,10)(H2,6,7,8);;;/q;3*+1/p-3

InChI key

ZJGSIAARCDPUSN-UHFFFAOYSA-K

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Intermediate in terpene biosynthesis

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Analysis Note

NMR-Internal Standard: KHP
Assay (quantitative NMR) ≥80.0% (calculated as free acid)

Other Notes

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
384261953344722
application(s)

clinical testing

application(s)

clinical testing

application(s)

food and beverages

application(s)

clinical testing

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

grade

analytical standard

grade

analytical standard

grade

analytical standard

grade

analytical standard

assay

≥95.0% (TLC)

assay

≥95.0% (TLC)

assay

≥95.0% (HPLC)

assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label


Still not finding the right product?


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters