Skip to Content
MilliporeSigma

39302

Okadaic acid solution

25 μg/g in methanol (nominal concentration), certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

35-Demethyl-DTX 1 solution, 35-Demethyldinophysistoxin 1 solution, NSC 677083

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C44H68O13
CAS Number:
Molecular Weight:
805.00
UNSPSC Code:
41116107
NACRES:
NA.24
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

grade

certified reference material, TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

concentration

25 μg/g in methanol (nominal concentration)

format

single component solution

storage temp.

−20°C

SMILES string

O1[C@]7(O[C@@H](CC[C@H]7O)C[C@](O)(C)C(=O)O)C=C(C[C@H]1[C@H](C)\C=C\[C@@H]2O[C@@]3(OC4[C@H](O[C@@H](C(=C)[C@H]4O)[C@@H](O)C[C@@H](C5O[C@]6(OCCCC6)CC[C@H]5C)C)CC3)CC2)C

InChI

1S/C44H68O13/c1-25-21-34(55-44(23-25)35(46)12-11-31(54-44)24-41(6,50)40(48)49)26(2)9-10-30-14-18-43(53-30)19-15-33-39(57-43)36(47)29(5)38(52-33)32(45)22-28(4)37-27(3)13-17-42(56-37)16-7-8-20-51-42/h9-10,23,26-28,30-39,45-47,50H,5,7-8,11-22,24H2,1-4,6H3,(H,48,49)/b10-9+/t26-,27-,28+,30+,31+,32+,33-,34+,35-,36-,37?,38+,39?,41-,42+,43-,44-/m1/s1

InChI key

QNDVLZJODHBUFM-AAWJMCDUSA-N

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to the SI through a primary reference material from a NMI.
Certified content incl. uncertainty and expiry date are stated on the enclosed certificate.

For the analysis of marine toxins in seafood.

Check out our complete range of marine toxins reference materials

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library