Skip to Content
MilliporeSigma

Skip To

36484

2-Stearoylglycerol

analytical standard

Synonym(s):

β-Monostearin, 2-Monostearin, 2-Monostearoylglycerol, 2-Octadecanoylglycerol, Glycerol β-monostearate, Octadecanoic acid 2-hydroxy-1-(hydroxymethyl)ethyl ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C21H42O4
CAS Number:
Molecular Weight:
358.56
UNSPSC Code:
85151701
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
1799576
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic

Quality Level

grade

analytical standard

assay

≥95% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

functional group

ester

storage temp.

−70°C

SMILES string

O=C(CCCCCCCCCCCCCCCCC)OC(CO)CO

InChI

1S/C21H42O4/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-21(24)25-20(18-22)19-23/h20,22-23H,2-19H2,1H3

InChI key

YQEMORVAKMFKLG-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
2701217561441679
grade

analytical standard

grade

-

grade

analytical standard

grade

analytical standard

format

neat

format

-

format

neat

format

neat

Quality Level

100

Quality Level

200

Quality Level

100

Quality Level

100

application(s)

food and beverages

application(s)

-

application(s)

food and beverages

application(s)

food and beverages

shelf life

limited shelf life, expiry date on the label

shelf life

-

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

storage temp.

−70°C

storage temp.

-

storage temp.

−70°C

storage temp.

−20°C


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



B C Mortimer et al.
Biochimica et biophysica acta, 1046(1), 46-56 (1990-08-28)
In rats, remnant particles derived from chylomicron-like emulsions containing 1,3-dioleoyl-2-stearoylglycerol (OSO) are removed from plasma more slowly than remnants derived from triolein emulsions. The effect associated with a saturated acyl chain at the glycerol 2-position could be reproduced by incorporating
Qi-Zhi Su et al.
Talanta, 202, 285-296 (2019-06-07)
Atmospheric pressure gas chromatography (APGC) coupled to quadrupole time-of-flight (QTOF) and electron ionization mass spectrometry together with commercial library search are two complementary techniques for non-target screening of volatile and semi-volatile compounds. Optimization was first conducted to achieve easier search



Global Trade Item Number

SKUGTIN
36484-25MG04061831825589

Questions

Reviews

No rating value

Active Filters