Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

36323

Sigma-Aldrich

(S)-6-Oxo-2-piperidinecarboxylic acid

≥95.0% (HPLC)

Synonym(s):

6-Oxo-L-pipecolic acid, (S)-2-Piperidone-6-carboxylic acid, L-Pyrohomoglutamic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H9NO3
CAS Number:
Molecular Weight:
143.14
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95.0% (HPLC)

impurities

≤5.0% water

functional group

carboxylic acid

SMILES string

OC(=O)[C@@H]1CCCC(=O)N1

InChI

1S/C6H9NO3/c8-5-3-1-2-4(7-5)6(9)10/h4H,1-3H2,(H,7,8)(H,9,10)/t4-/m0/s1

InChI key

FZXCPFJMYOQZCA-BYPYZUCNSA-N

Application

(S)-6-Oxo-2-piperidinecarboxylic acid can be used as a reactant to synthesize:
  • Functionalized β-lactam N-heterocycles via carboxymethylproline synthase catalyzed cyclization reactions. Pro-(S)-C5 branched [4.3.1] aza-amide bicycles as potential inhibitors for FK506-binding proteins.
  • Ethyl (S)-2-(6-oxopiperidin-2-yl)acetate, which is reduced using LiBH4 to produce optically pure hydroxymethyl lactams.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carboxymethylproline synthase catalysed syntheses of functionalised N-heterocycles
Hamed RB, et al.
Chemical Communications (Cambridge, England), 46(9), 1413-1415 (2010)
Preparation of optically pure ω-hydroxymethyl lactams
Huang S-B, et al.
Synthetic Communications, 19(20), 3485-3496 (1989)
Rational Design and Asymmetric Synthesis of Potent and Neurotrophic Ligands for FK506-Binding Proteins (FKBPs)
Pomplun S, et al.
Angewandte Chemie (International ed. in English), 54(1), 345-348 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service