Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

34760BC

Millipore

Assurance® GDS E. coli O157:H7 Positive Control

For use with Assurance® GDS E. coli O157:H7 Tq

Synonym(s):

Positive Control for PCR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116135
NACRES:
NA.85

Product Name

GDS Positive Control for E. coli O157:H7, BioControl, For use with GDS E. coli O157:H7

Quality Level

form

liquid

shelf life

12 mo.

packaging

vial of 5 mL (sufficient for 166 tests)

manufacturer/tradename

BioControl Systems 34760-250

storage condition

(Tightly closed)

technique(s)

PCR: suitable

application(s)

food and beverages
pathogen testing

compatibility

for use with BioControl GDS

storage temp.

2-8°C

suitability

Escherichia coli (O157:H7)

General description

GDS Positive Control for E. coli O157:H7 is used with the GDS E. coli O157:H7 Tq assay, which is an automated nucleic acid amplification system for the detection of pathogenic E. coli O157:H7 in foods, ingredients, and environmental samples.

Application

GDS Positive Control for E. coli O157:H7 is used for confirmation of E.coli O157:H7 detection in Assurance® GDS assays.

Components

One vial, 5 mL, sufficient for 166 tests

Testing Method

PCR

Legal Information

Assurance is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service