Skip to Content
MilliporeSigma

34745BC

Assurance® GDS Listeria Resuspension Buffer Tq PCR Reagent

For use with Assurance® GDS Listeria Tq

Synonym(s):

Buffer for PCR Sample Prep

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41106306
NACRES:
NA.85
Suitability:
Listeria spp.
Manufacturer/tradename:
BioControl Systems 34745-100C
Application(s):
food and beverages
pathogen testing
Technique(s):
PCR: suitable
Storage temp.:
2-8°C
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

GDS Listeria Resuspension Buffer Tq, BioControl, For use with GDS Listeria

Quality Segment

form

liquid

shelf life

24 mo.

packaging

bottle of 70 mL

manufacturer/tradename

BioControl Systems 34745-100C

storage condition

(Tightlyclosed in a dry, cool, and well-ventilated place)

technique(s)

PCR: suitable

application(s)

food and beverages
pathogen testing

compatibility

for use with BioControl GDS

storage temp.

2-8°C

suitability

Listeria spp.

General description

GDS Listeria Resuspension Buffer Tq is one of the components of the Assurance® GDS Listeria monocytogenes Tq, which is a genetic detection system for Listeria monocytogenes. It is an automated nucleic acid amplification system for the detection of Listeria monocytogenes from food and environmental samples. This larger size format is commonly purchased with Listeria automation kits for use with Assurance® GDS PickPen® PipetMax® (PPMX).

Application

GDS Listeria Resuspension Buffer Tq is used during sample preparation for the detection of Listeria monocytogenes from food and environmental samples.
PCR

Other Notes

One bottle, 70 mL

Legal Information

Assurance is a registered trademark of Merck KGaA, Darmstadt, Germany
PICKPEN is a registered trademark of Merck KGaA, Darmstadt, Germany
PIPETMAX is a registered trademark of Gilson, Inc




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library