Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

34237

Supelco

Cinmethylin

PESTANAL®, analytical standard

Synonym(s):

exo-1-Methyl-4-(1-methylethyl)-2-[(2- methylphenyl)methoxy]-7-oxabicyclo[2.2.1]heptane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H26O2
CAS Number:
Molecular Weight:
274.40
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

description

mixture of isomers

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CC(C)[C@]12CC[C@](C)(O1)[C@@H](C2)OCc3ccccc3C

InChI

1S/C18H26O2/c1-13(2)18-10-9-17(4,20-18)16(11-18)19-12-15-8-6-5-7-14(15)3/h5-8,13,16H,9-12H2,1-4H3/t16-,17+,18-/m1/s1

InChI key

QMTNOLKHSWIQBE-FGTMMUONSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cinmethylin belongs to the cineole family and is a pre-emergence herbicide developed for the control of annual grass weeds.

Application

Cinmethylin may be used as an analytical reference standard for the determination of the analyte in food matrices, fruits and vegetables by various chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

296.6 °F

Flash Point(C)

147 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multiresidue determination of 375 organic contaminants including pesticides, polychlorinated biphenyls and polyaromatic hydrocarbons in fruits and vegetables by gas chromatography--triple quadrupole mass spectrometry with introduction of semi-quantification approach.
Banerjee K, et al.
Journal of Chromatography A, 1270(2), 283-295 (2012)
Rapid multiresidue method for the determination of more than 300 pesticide residues in food.
Okihashi M, et al.
Food, 1(2), 101-110 (2007)
The physical and chemical properties of the herbicide cinmethylin (SD 95481).
Grayson BT, et al.
Pestic. Sci., 21(2), 143-153 (1987)
Roberto Busi et al.
Pest management science, 76(8), 2601-2608 (2020-02-23)
Multiple-herbicide resistance in Lolium rigidum and other weed species is increasingly exerting pressure on herbicide discovery research for solutions against resistance-prone weeds. In this study we investigate: (i) the responses of L. rigidum populations and wheat to the new herbicide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service