Skip to Content

34142

Fumonisin B2 solution

~50 μg/mL in acetonitrile: water (50:50), analytical standard

Synonym(s):

FB2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C34H59NO14
CAS Number:
Molecular Weight:
705.83
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

shelf life

limited shelf life, expiry date on the label

concentration

~50 μg/mL in acetonitrile: water (50:50)

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

single component solution

storage temp.

−20°C

SMILES string

CCCC[C@@H](C)[C@@H](OC(=O)C[C@@H](CC(O)=O)C(O)=O)[C@H](C[C@@H](C)CCCCCC[C@@H](O)C[C@H](O)[C@H](C)N)OC(=O)C[C@@H](CC(O)=O)C(O)=O

InChI

1S/C34H59NO14/c1-5-6-12-21(3)32(49-31(43)18-24(34(46)47)16-29(40)41)27(48-30(42)17-23(33(44)45)15-28(38)39)14-20(2)11-9-7-8-10-13-25(36)19-26(37)22(4)35/h20-27,32,36-37H,5-19,35H2,1-4H3,(H,38,39)(H,40,41)(H,44,45)(H,46,47)

InChI key

UXDPXZQHTDAXOZ-UHFFFAOYSA-N

General description

Fumonisin B2 belongs to the family of toxins known as fumonisins. It is produced by several species of Fusarium molds primarily by Fusarium verticillioide. It is one of the major mycotoxin contaminant in corn and corn-based foods. Fumonisin B2 is a structural analog of fumonisin B1 but it is more cytotoxic than the latter and it inhibits sphinganine-N-acetyltransferase (ceramide synthase).
Fumonisin B2 is a carcinogenic mycotoxin generally present on corn based food and feed-stuff, which is produced by the fungus Fusarium verticillioides and Fusarium moniliforme. Fumonisin B2 can also be detected in the industrially important fungi, Aspergillus niger.
Certan Vial

Application

Fumosin B2 maybe used to study the effect of temperatures from ambient to 150°C and solvent compositions (methanol-water, acetonitrile-methanol-water, ethanol-water, and 100% water) on recovery of fumonisins from corn products.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analysis Note

purity : ≥94.0% (HPLC)


Still not finding the right product?

Explore all of our products under Fumonisin B2 solution


pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

flash_point_f

35.6 °F

flash_point_c

2 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library