Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

33965

Supelco

Fenamidone

PESTANAL®, analytical standard

Synonym(s):

(S)-1-Anilino-4-methyl-2-methylthio-4-phenyl-2-imidazolin-5-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H17N3OS
CAS Number:
Molecular Weight:
311.40
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CSC1=N[C@](C)(C(=O)N1Nc2ccccc2)c3ccccc3

InChI

1S/C17H17N3OS/c1-17(13-9-5-3-6-10-13)15(21)20(16(18-17)22-2)19-14-11-7-4-8-12-14/h3-12,19H,1-2H3/t17-/m0/s1

InChI key

LMVPQMGRYSRMIW-KRWDZBQOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Fenamidone is an imidazolinone fungicide. It is effective in treating downy mildew (Plasmopara viticola) in grapevines.

Application

Fenamidone may be used as a reference standard for the determination of fenamidone in vegetable samples by gas chromatography (GC).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Validation and global uncertainty of a gas chromatographic with mass spectrometry method for fenamidone analysis in grapes and wines.
de Melo Abreu S, et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 42(7), 817-822 (2007)
Yengkhom Bijen Kumar et al.
Journal of food science and technology, 57(6), 2061-2069 (2020-05-21)
Dissipation behavior and degradation kinetics of fenamidone + mancozeb (Sectin 60 WG) and iprovalicarb + propineb (Melody Duo 66.75 WP) in tomato were studied at recommended dose (RD) and double dose (DD) of application. The analysis of the field samples were carried out by
Nikhilesh Dhar et al.
Plant disease, 104(1), 82-93 (2019-11-19)
The lettuce downy mildew pathogen, Bremia lactucae, is an obligate oomycete that causes extensive produce losses. Initial chlorotic symptoms that severely reduce the market value of the produce are followed by the appearance of white, downy sporulation on the abaxial
Alberto Angioni et al.
Pest management science, 68(4), 543-547 (2011-11-22)
Greenhouse tomato production is intended exclusively for fresh consumption. Typical greenhouse conditions provide a conducive environment for the development of different fungi, principally late blight and grey mould, that can destroy plants and fruits. For this reason, different fungicides with
Milica S Jovetić et al.
Arhiv za higijenu rada i toksikologiju, 69(3), 264-274 (2018-10-05)
To contribute to the development of urban beekeeping, we designed this study to obtain more information about the contamination of urban bee products with toxic metals, polycyclic aromatic hydrocarbons (PAHs), and pesticides. The samples of honey (N=23), pollen (N=13), and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service