Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

33452

Supelco

Parathion-ethyl-d10

PESTANAL®, analytical standard

Synonym(s):

O,O-Diethyl-d10 O-(4-nitrophenyl) phosphorothioate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10D10H4NO5PS
CAS Number:
Molecular Weight:
301.32
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

mass shift

M+10

storage temp.

2-8°C

SMILES string

[2H]C([2H])([2H])C([2H])([2H])OP(=S)(Oc1ccc(cc1)[N+]([O-])=O)OC([2H])([2H])C([2H])([2H])[2H]

InChI

1S/C10H14NO5PS/c1-3-14-17(18,15-4-2)16-10-7-5-9(6-8-10)11(12)13/h5-8H,3-4H2,1-2H3/i1D3,2D3,3D2,4D2

InChI key

LCCNCVORNKJIRZ-MWUKXHIBSA-N

Application

It may be used as an internal standard for the determination of parathion-ethyl in grape seed extract using gas chromatography coupled to tandem mass spectrometry with a triple quadrupole analyzer (GC-QqQ-MS/MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Inhalation - Acute Tox. 1 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

248.0 °F - closed cup

Flash Point(C)

120 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multi-pesticide residue analysis in nutraceuticals from grape seed extracts by gas chromatography coupled to triple quadrupole mass spectrometry
Nieto-Garcia, A.J., et al.
Food Control, 47, 369-380 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service