Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

32834

Supelco

Leucocrystal Violet-d6

analytical standard

Synonym(s):

Bis(4-dimethylaminophenyl)-4-dimethylamino-d6-phenylmethane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25D6H25N3
CAS Number:
Molecular Weight:
379.57
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

mass shift

M+6

SMILES string

[2H]C([2H])([2H])N(c1ccc(cc1)C(c2ccc(cc2)N(C)C)c3ccc(cc3)N(C)C)C([2H])([2H])[2H]

InChI

1S/C25H31N3/c1-26(2)22-13-7-19(8-14-22)25(20-9-15-23(16-10-20)27(3)4)21-11-17-24(18-12-21)28(5)6/h7-18,25H,1-6H3/i1D3,2D3

InChI key

OAZWDJGLIYNYMU-WFGJKAKNSA-N

General description

Leucocrystal Violet-d6 is a deuterated analog of leucocrystal violet dye.

Application

Leucocrystal Violet-d6 may be used as an internal standard for the determination of leucocrystal violet in aquaculture products by liquid chromatography-tandem mass spectrometry (LC-MS/MS).
Leucocrystal violet-d6 may be used as an internal standard in the determination of triphenylmethane dyes in rainbow trout samples using liquid chromatography coupled with tandem mass spectrometry (LC-MS/MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

HPLC-q/LIT-MS Determination of Malachite Green, Crystal Violet and their Metabolites in Aquatic Products and Feed.
ZHAO SZ, et al.
Physical Testing and Chemical Analysis Part B:Chemical Analysis, 1349(1), 9-9 (2014)
A rapid and simple method for simultaneous determination of triphenylmethane dye residues in rainbow trouts by liquid chromatography-tandem mass spectrometry.
Kaplan M, et al.
Journal of Chromatography A, 1349(12), 37-43 (2014)
Muammer Kaplan et al.
Journal of chromatography. A, 1349, 37-43 (2014-05-29)
A rapid and simple LC-MS/MS method was developed and optimized for screening and confirmation of triphenylmethane dyes including malachite green (MG), leucomalachite green (LMG), crystal violet (CV), leucocrystal violet (LCV) and brilliant green (BG) in fish muscle with skin. Leucocrystal
D Hurtaud-Pessel et al.
Journal of chromatography. A, 1218(12), 1632-1645 (2011-02-12)
A method is described for the identification and the quantitative determination of the triphenylmethane dyes, malachite green (MG), crystal violet (CV), brilliant green (BG) and leuco malachite green (LMG) and leuco crystal violet (LCV). The analytes were isolated from the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service