Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

32736

Supelco

Carprofen-d3

VETRANAL®, analytical standard

Synonym(s):

6-Chloro-α-methyl-d3-9H-carbazole-2-acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15D3H9ClNO2
CAS Number:
Molecular Weight:
276.73
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

mass shift

M+3

SMILES string

[2H]C([2H])([2H])C(C(O)=O)c1ccc2c(c1)[nH]c3ccc(Cl)cc23

InChI

1S/C15H12ClNO2/c1-8(15(18)19)9-2-4-11-12-7-10(16)3-5-13(12)17-14(11)6-9/h2-8,17H,1H3,(H,18,19)/i1D3

InChI key

PUXBGTOOZJQSKH-FIBGUPNXSA-N

General description

Carprofen-d3 is the deuterated analog of the nonsteroidal anti-inflammatory drug, carprofen.

Application

Carprofen-d3 may be used as an analytical standard for the determination of carprofen in animal muscles and milk samples by liquid chromatography–tandem mass spectrometry (LC-MS/MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of non-steroidal anti-inflammatory drugs and their metabolites in milk by liquid chromatography-tandem mass spectrometry.
Jedziniak P, et al.
Analytical and Bioanalytical Chemistry, 403(10), 2955-2963 (2012)
Piotr Jedziniak et al.
Analytica chimica acta, 672(1-2), 85-92 (2010-06-29)
A confirmatory method for the determination of residues of nine non-steroidal anti-inflammatory drugs and one metabolite in animal muscles has been developed. After enzymatic hydrolysis samples were extracted with acetonitrile and cleaned up using alumina and C(18) SPE cartridges. Liquid
Simultaneous determination of residues of non-steroidal anti-inflammatory drugs and glucocorticosteroids in animal muscle by liquid chromatography-tandem mass spectrometry.
Jedziniak P, et al.
Food Analytical Methods, 9(6), 1837-1848 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service