Skip to Content
MilliporeSigma

32609

Sterigmatocystin

Synonym(s):

3a,12c-Dihydro-8-hydroxy-6-methoxy-7H-furol[3′,2′:4,5]furo[2,3-c]xanthen-7-one

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C18H12O6
CAS Number:
Molecular Weight:
324.28
UNSPSC Code:
85151701
NACRES:
NA.24
PubChem Substance ID:
EC Number:
233-158-6
Beilstein/REAXYS Number:
53259
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

grade

analytical standard, reference material

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

−20°C

SMILES string

COc1cc2OC3OC=CC3c2c4Oc5cccc(O)c5C(=O)c14

InChI

1S/C18H12O6/c1-21-11-7-12-13(8-5-6-22-18(8)24-12)17-15(11)16(20)14-9(19)3-2-4-10(14)23-17/h2-8,18-19H,1H3

InChI key

UTSVPXMQSFGQTM-UHFFFAOYSA-N

General description

Sterigmatocystin is classified under the mycotoxin family of compounds that is usually produced by fungi of several species of Aspergillus.

Application

Sterigmatocystin may be used as an analytical reference standard for the determination of the analyte in:
  • Grain samples by high-performance liquid chromatography-electrospray positive ionization-tandem mass spectrometry (HPLC-ESI+-MS/MS).
  • Sweet pepper samples by LC-ESI+-MS/MS.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Initiates lung and liver tumors under experimental conditions.


Still not finding the right product?

Explore all of our products under Sterigmatocystin


pictograms

Skull and crossbonesHealth hazard

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral - Carc. 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library